Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Germaine™ Laboratories AimStep™ H. Pylori

Catalog No. 23111395
Change view
Click to view available options
Clia Complexity:
Waived (Whole Blood)
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Clia Complexity
23-111-395 Waived (Whole Blood)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 23-111-395 Supplier Germaine™ Laboratories Supplier No. 33070
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

AimStep™ H. Pylori is a rapid cassette test for the qualitative detection of Helicobacter pylori in serum, plasma, or whole blood. Results in 10 minutes. CLIA Waived for whole blood.

  • 91.6% agreement with biopsy histology
  • Single reagent procedure
  • CAP/NCCLS formatted procedures available via email or fax

Specifications

Content And Storage Ambient
CE Marker No
Clia Complexity Waived (Whole Blood)
Detectable Analytes H. pylori
Sample Type Serum, Plasma, or Whole Blood
Includes Controls, Capillary Tubes, Buffer, Specimen Dropper
Quantity 20 Tests
Test Time 10 min.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.