Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Germaine™ Laboratories AimStep™ Mono Rapid Cassette Test

Catalog No. 23111302
Change view
Click to view available options
Quantity:
25 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-111-302 25 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 23-111-302 Supplier Germaine™ Laboratories Supplier No. 77025
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Germaine™ Laboratories AimStep™ Mono Rapid Cassette Test is for the qualitative detection of Infectious Mononucleosis (IM) heterophile antibodies in serum, plasma, or whole blood

  • Higher sensitivity to meet your needs
  • Positive and negative controls, capillary tubes, buffer and specimen dropper included
  • Priced to meet your budget requirements
  • CAP/NCCLS formatted procedures available via email or fax; just add your name
  • CLIA Waived for whole blood
  • Results in 5 minutes

Specifications

Content And Storage 2°C to 30°C
CPT Code 86308QW
Clia Complexity Waived
Detectable Analytes Mononucleosis Antibodies
Sample Type Serum, plasma, or whole blood
Disposable Single-use
Quantity 25 Tests
Test Time 5 min.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.