Learn More
Germaine™ Laboratories AimStep Ovulation Test
Catalog No. 23111379
Change view
Click to view available options
Quantity:
30 Tests/Kit
Catalog No. Quantity
23-111-379 30 Tests/Kit
1 options
This item is not returnable. View return policy
Catalog No. 23-111-379 Supplier Germaine™ Laboratories Supplier No. 75030
Add to Cart
Add to Cart
This item is not returnable. View return policy
Qualitative Test to Detect the LH Surge (Luteinizing Hormone or hLH) in Urine to Aid in the Prediction of Ovulation
Results in 5 minutes
Specifications
Content And Storage 15°C to 30°C
Detectable Analytes Luteinizing Hormone Surge (LH or hLH)
Type Urine
Includes Test Devices; Pipettes
Packaging Type Individually wrapped foil pouches
Quantity 30 Tests/Kit