Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Germaine™ Laboratories AimStrip™ Pregnancy Test

Catalog No. 23111296
Change view
Click to view available options
Quantity:
30 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-111-296 30 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 23-111-296 Supplier Germaine™ Laboratories Supplier No. 88530
Add to Cart
Add to Cart

The Germaine™ Laboratories AimStrip™ Pregnancy Test is a dipstick method for the rapid qualitative determination of human chorionic gonadotropin (hCG) in urine to aid in the early detection of pregnancy.

  • DipStick format with built-in quality control check
  • Just dip into urine for 5 seconds and get results in 3 minutes or less
  • Room Temperature Storage
  • Individually foil-wrapped tests

Specifications

Certifications/Compliance CLIA Waived
Content And Storage 15°C to 30°C
CPT Code 81025QW
CE Marker Yes
Clia Complexity Waived
Detectable Analytes hCG
Sample Type Urine
Sensitivity 20 mIU/mL
Type Urine DipStick
Disposable Single-use
Quantity 30 Tests
Test Time 3 min.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.