Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Germaine™ Laboratories AimStep™ Pregnancy Test

Catalog No. 23111297
Change view
Click to view available options
Quantity:
30 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-111-297 30 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 23-111-297 Supplier Germaine™ Laboratories Supplier No. 97030
Add to Cart
Add to Cart

Germaine™ Laboratories AimStep™ Pregnancy Test is for the rapid qualitative determination of human chorionic gonadotropin (hCG) in urine to aid in the early detection of pregnancy. This test is for professional use.

  • CLIA Complexity: Waived
  • Test results within three minutes
  • Combination of monoclonal and polyclonal antibody reagent selectively detects elevated levels of human chorionic gonadotropin (hCG)
  • Positive specimens react with the specific antibody-hCG colored conjugate and form a colored line at the T (test) area
  • Absence of this colored line suggests a negative result
  • A colored line at the C (control) area will always appear regardless of the presence or absence of hCG

Specifications

Content And Storage 15°C to 30°C
CPT Code 81025QW
Clia Complexity Waived
Detectable Analytes hCG
Sample Type Urine
Sensitivity 20 mIU/mL
Type Urine Cassette Test
Disposable Single-use
Quantity 30 Tests
Test Time 3 min.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.