Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Germaine™ Laboratories AimStrip™ Adulteration Pads

Catalog No. 23111360
Change view
Click to view available options
Quantity:
25 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-111-360 25 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 23-111-360 Supplier Germaine™ Laboratories Supplier No. 40007
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Germaine™ Laboratories AimStrip™ Adulteration Pads are a semi-quantitative, color comparison screen for the detection of creatinine, nitrite, glutaraldehyde, pH, specific gravity, and oxidants-pyridinium chlorochromate (PCC) in urine

Specifications

Content And Storage 15°C to 30°C
Detectable Analytes Creatinine, Nitrite, Gluteraldehyde, pH, Specific Gravity, Oxidant, PCC
Type Urine Dipstick
Disposable Single-use
For Use With (Application) DOA
Quantity 25 Tests
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.