Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Germaine™ Laboratories AimStrip™ Dipstick Reagent Strips

Catalog No. 23111292
Change view
Click to view available options
Detectable Analytes:
Glucose
Ketone
Sensitivity:
5 to 10 mg/dL Acetoacetic Acid
50 to 100 mg/dL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Detectable Analytes Sensitivity
23-111-292 Glucose 50 to 100 mg/dL
23-111-291 Ketone 5 to 10 mg/dL Acetoacetic Acid
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 23-111-292 Supplier Germaine™ Laboratories Supplier No. 50100
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Germaine™ Laboratories AimStrip™ Dipstick Reagent Strips are for qualitative detection of analytes in urine

Specifications

Content And Storage 2°C to 30°C
Clia Complexity Waived
Detectable Analytes Glucose
Sample Type Urine
Sensitivity 50 to 100 mg/dL
Type Urinalysis Dipstick
Disposable Single-use
For Use With (Application) Urinalysis
Quantity 100 Tests
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.