Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Germaine™ Laboratories AimStrip™ Dipstick Reagent Strips for 5 Analytes

Catalog No. 23111294
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-111-294 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 23-111-294 Supplier Germaine™ Laboratories Supplier No. 55100
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Germaine™ Laboratories AimStrip™ Dipstick Reagent Strips for 5 Analytes is for the qualitative detection in urine samples

  • Directional indicator on bottle color chart reduces likehood of misaligning strip when reading results
  • Vertical color chart reads up an down the bottle, reduces rotating bottle while reading results
  • CAP/NCCLS formatted procedures available via email or fax; just add yur name
  • May be read visually or on optical CLIA waived AimStrip™ Urine Analyzer

Specifications

Content And Storage 2°C to 30°C
Clia Complexity Waived
Detectable Analytes Glucose, Protein, Ketone, pH, Blood
Sample Type Urine
Disposable Single-use
For Use With (Application) Urinalysis
Quantity 100 Tests
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.