Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Germaine Laboratories, Inc. AimStrip Fentanyl Diptstick

Catalog No. 23111419
Change view
Click to view available options
Sensitivity:
10 ng/mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Sensitivity
23-111-419 10 ng/mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 23-111-419 Supplier Germaine Laboratories, Inc. Supplier No. 43345
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Fentanyl Test Strips-for liquids and powders, detects levels as low as. 10 or 20 ng/mL. Almost no cross reactivity with Methamphetamine or MDMA.

  • AimStrip Fentanyl Test Strips-for liquids and powders.
  • 25 Strips/Kit. 10 ng/mL.
  • Ultra-High Sensitivity-can detect Fentanyl down to low levels of 10 or 20 ng/mL.
  • Exceptional Specificity- almost no cross reactivity with Methamphetamine or MDMA.
  • For forensic use only.
  • Results in 5 minutes.
  • User Friendly Kit - Simply immerse the dipstick in the liquid for 15 seconds and read the results at 5 minutes. Do not read results after 10 Minutes. Do not let the solution touch the MAX line as this would cause an inconclusive result.
  • FEN Test Strips for Testing Drugs in Urine Sample or Pills/Powders in Liquid. For testing a Powder, add 1 mL water to 50 mg substance and mix well. For testing a Pill, crush/scrape off 50 mg of the pill and add 1 mL water. Dip the strip for 15 seconds in the mixture/liquid that is to be tested.
  • Made in USA

Specifications

Description For Forensic Use Only
Clia Complexity Not Applicable
Detectable Analytes Fentanyl (FEN)
Format Test Strips, Kit
Sample Type Liquids and Powders
Sensitivity 10 ng/mL
Type Fentanyl Diptstick
Disposable Single-use
Includes 25 DipSticks per Kit, package insert.
Packaging Type Test Kit
Test Time 5 min.
Analysis Time 5 min.
Form Ready-to-Use (RTU)
Show More Show Less

For forensic use only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.