Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Germaine™ Laboratories AimStrip MACRE Strips

Catalog No. 23111347
Change view
Click to view available options
Quantity:
25 Strips
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-111-347 25 Strips
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 23-111-347 Supplier Germaine™ Laboratories Supplier No. 52275
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Germaine™ Laboratories AimStrip™ MACRE Strips are dipstick reagent strips for the qualitative detection of Microalbumin and Creatinine

  • Detects Microalbumin & Creatinine as low as 10mg/L
  • Sensitivity as low as 2.0 mg/dL, CLIA Waived
  • Semi-Quantitative A:C ratio results in one minute
  • Exclusively read on AimStrip™ Urine Analyzer 2
  • Same procedure as leading national brand

Specifications

Content And Storage 2°C to 30°C
CPT Code Albumin 82044QW, Creatine 82570QW
CE Marker Yes
Clia Complexity Waived
Detectable Analytes Microalbumin, Creatinine
Sample Type Urine
Disposable Single-use
For Use With (Application) Urinalysis
Quantity 25 Strips
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.