Learn More
Germaine™ Laboratories AimStrip™ Tandem Glucose Control Solution
Catalog No. 23111387
Change view
Click to view available options
Level:
Tri-Level Control Solution
Catalog No. Level
23-111-387 Tri-Level Control Solution
1 options
This item is not returnable. View return policy
Catalog No. 23-111-387 Supplier Germaine™ Laboratories Supplier No. 77512
Add to Cart
Add to Cart
This item is not returnable. View return policy
Used to test the precision of the AimStrip™ Tandem Glucose Measuring System and to detect systematic analytic deviations that may arise from reagent or analytical variation.
  • Control Solution for AimStrip™ Tandem Blood Glucose Test Strips
  • Simply insert test, apply specimen & read results
Specifications
Detectable Analytes Glucose
For Use With (Equipment) AimStrip™ TANDEM Measuring System
Includes Contol Solution Level 1 (3.5 mL), Contol Solution Level 2 (3.5 mL), Contol Solution Level 3 (3.5 mL), 10 Capillary Rods
Level Tri-Level Control Solution
Quantity 3 x 3.5 mL