Learn More
Germaine™ Laboratories AimStrip™ Tandem Lipid Profile & Glucose Starter Bundle
Catalog No. 23111389
Change view
Click to view available options
Analysis Time:
3 sec. to 2 min.
Catalog No. Analysis Time
23-111-389 3 sec. to 2 min.
1 options
This item is not returnable. View return policy
Catalog No. 23-111-389 Supplier Germaine™ Laboratories Supplier No. 77345
Add to Cart
Add to Cart
This item is not returnable. View return policy
Bundle includes 1 meter and 1 pack of test strips for Total Cholesterol (TC) and Blood Glucose (BG).
  • Tests Cholesterol, Triglycerides, and Blood Glucose all in ONE Meter
  • Zero Maintenance - No servicing of meter is required
  • Meter has a 1-year warranty
  • Battery Operated
  • Handheld Meter for POC Testing
  • Easy to read LCD Results
  • Simply insert test, apply specimen & read results
  • No specimen preparation required
Specifications
Sample Type Capillary Whole Blood
Description Tests for Cholesterol, Blood Glucose, and Triglycerides all in ONE Meter
Analysis Time 3 sec. to 2 min.
Includes 1 Meter, 1 Box of Total Cholesterol Strips, 1 Box of Triglycerides Strips
Electrical Requirements Battery Operated - 2 AAA Batteries
Memory Stores 200 Results