Learn More
Germaine™ Laboratories AimStrip™ Tandem Meter (Cholesterol, Triglycerides and Blood Glucose l)
Catalog No. 23111381
Change view
Click to view available options
Analysis Time:
3 sec. (Glucose), 2 min. (Cholesterol and Triglycerides)
Catalog No. Analysis Time
23-111-381 3 sec. (Glucose), 2 min. (Cholesterol and Triglycerides)
1 options
This item is not returnable. View return policy
Catalog No. 23-111-381 Supplier Germaine™ Laboratories Supplier No. 77301
Add to Cart
Add to Cart
This item is not returnable. View return policy
For testing Cholesterol, Triglycerides and Blood Glucose levels in capillary whole blood.
  • Tests Cholesterol, Triglycerides, and Blood Glucose all in ONE Meter
  • Zero Maintenance - No servicing of meter is required
  • Meter has a 1-year warranty
  • Battery Operated
  • Handheld Meter for POC Testing
  • Easy to read LCD Results
  • Simply insert test, apply specimen & read results
  • No specimen preparation required
Specifications
Analysis Time 3 sec. (Glucose), 2 min. (Cholesterol and Triglycerides)
Sample Size 0.3 to 5 μL
Includes 1 Meter, Operational Manual, Warranty Card (1 year), Carrying Case, 2 AAA Batteries, Log Book