Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Germaine™ Laboratories AimStrip™ Tandem Total Cholesterol Control Solution

Catalog No. 23111385
Change view
Click to view available options
Level:
Bi-level
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Level
23-111-385 Bi-level
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 23-111-385 Supplier Germaine™ Laboratories Supplier No. 77312
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Used to test the precision of the AimStrip™ Tandem Lipid Profile Measuring System and to detect systematic analytic deviations that may arise from reagent or analytical variation.

  • Control Solution for AimStrip™ Tandem Total Cholesterol Test Strips
  • Simply insert test, apply specimen & read results

Specifications

Content And Storage 8°C to 30°C
Detectable Analytes Cholesterol (Total)
For Use With (Equipment) AimStrip™ TANDEM Measuring System
Includes Contol Solution Level 1 (3 mL), Contol Solution Level 2 (3 mL), 10 Capillary Rods
Level Bi-level
Quantity 2 x 3 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.