Learn More
Germaine™ Laboratories AimStrip™ Tandem Total Cholesterol Test Strips
Catalog No. 23111382
Change view
Click to view available options
Calibration:
Plasma- Equivalent
Catalog No. Calibration
23-111-382 Plasma- Equivalent
1 options
This item is not returnable. View return policy
Catalog No. 23-111-382 Supplier Germaine™ Laboratories Supplier No. 77310
Add to Cart
Add to Cart
This item is not returnable. View return policy
Used to measure total cholesterol (TC) and is used in the diagnosis and treatment of lipid disorders (such as diabetes mellitus), atherosclerosis, and various liver and renal diseases.
  • Tests Cholesterol, Triglycerides, and Blood Glucose all in ONE Meter
  • Includes 10 Strips/Box and 10 Capillary Rods
  • Results in 2 minutes
  • Simply insert test, apply specimen & read results
  • No specimen preparation required
Specifications
Calibration Plasma- Equivalent
Content And Storage 2°C to 30°C
Clia Complexity Waived
Detectable Analytes Blood Glucose, Total Cholesterol, Triglycerides