Learn More
Germaine™ Laboratories AimStrip Urine Analyzer 2
Description
- Reads urine tests strips of 2 or more parameter combinations, up to 10 analytes
- Barcode reader, optional and can be purchased separately
- Languages include English, Spanish and Portuguese
- CLIA Waived
- Compatible with Fisherbrand 10-SG Reagent strips and Germaine AimStrip urine reagent tests strips
Specifications
Specifications
| Product Type | Urine Analyzer |
| Display Type | Digital |
| Languages | English, Spanish, Portuguese |
| Memory | 2000 results |
| Throughput | Single test mode: 60 tests/hr, Continuous Mode: 120 tests/hr |
| Dimensions (L x W x H) | 11 x 7.5 x 6 in. |
| Weight (English) | 4.5 lb. |
| For Use With (Application) | Semi-quantitative detection of analytes in urine. |
| Electrical Requirements | 100/240 VAC 50/60 Hz |
| Frequency | 50/60 Hz |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.