Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Germaine™ Laboratories AimTab™ Bluing Tablets

Catalog No. 23111362
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-111-362 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 23-111-362 Supplier Germaine™ Laboratories Supplier No. 40021
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Germaine™ Laboratories AimTab™ Bluing Tablets aid in leak detection by plumbers and prevent adulteration in all drug testing collections

  • Place tablet into toilet bowl before collection and turns water vibrant blue in less than a minute
  • Use one tablet per flush
  • Meets Federal DOT Regulation 49 CFR 40.41
  • Plumbers use for testing customers toilets for leaking flapper and ball valves by placing into toilet tank
  • CAP / CLSI formatted procedures available via fax or email - just add your name!
  • Store at Room Temperature

Specifications

Content And Storage 10°C to 35°C
Sample Type Water
Disposable Single-use
For Use With (Application) DOA Testing
Quantity 100 Tests
Test Time <1 min.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.