Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Germaine™ Laboratories AimTab™ Ketone Serum Controls

Catalog No. 23111367
Change view
Click to view available options
Quantity:
2 x 2 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-111-367 2 x 2 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 23-111-367 Supplier Germaine™ Laboratories Supplier No. 13112
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Germaine™ Laboratories Ketone Serum Controlsare for testing Ketone tablet tests

Negative control (1 x 2mL), Positive control (1 x 2mL)

Specifications

Content And Storage 2°C to 8°C
Detectable Analytes Ketones
For Use With (Equipment) Ketone tablet tests
Quantity 2 x 2 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.