Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Germaine™ Laboratories AimTab™ Ketone Tablets

Catalog No. 23111357
Change view
Click to view available options
Quantity:
100 Tests/Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-111-357 100 Tests/Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 23-111-357 Supplier Germaine™ Laboratories Supplier No. 13100
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Includes

100 Ketone Tablets, Package Insert

For the semi-quantitative determination of Ketones (acetoacetic acid & acetone) in urine, serum, plasma and whole blood. Replaces the discontinued Bayer Acetest. Same test procedure at the Acetest™. Detects as little as 5mg/dl of acetoacetic acid in urine.

  • Approximately 10 times more sensitive to acetoacetic acid than acetone
  • Specific for acetoacetic acid and acetone and will not react with betahydroxybutyric acid
  • CAP / CLSI formatted procedure available electronically
  • Manufactured in the U.S.A.
  • Positive Result: Purple color

Warranty and Services

18 months

Specifications

Content And Storage Room Temperature
Clia Complexity Moderate (Serum, Plasma or Whole Blood), Waived (Other)
Type Ketone Tablets
For Use With (Application) Laboratory diagnostic testing
Includes 100 Ketone Tablets, Package Insert
Quantity 100 Tests/Bottle
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.