Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Germaine™ Laboratories AimTab™ Reducing Substances Tablets

Catalog No. 23111364
Change view
Click to view available options
Sample Type:
Urine, Wine, Stool, Other Specimens
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Sample Type
23-111-364 Urine, Wine, Stool, Other Specimens
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 23-111-364 Supplier Germaine™ Laboratories Supplier No. 13320
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Germaine™ Laboratories AimTab™ Reducing Substances Tablets are for the semi-quantitative determination for the amount of reducing substances (generally glucose) in urine and other fluids

  • Quick & easy results for testing urine, stool, wine and other liquids.
  • Detects as little as 0.25 g/dL of glucose.
  • Detects glucose and other reducing sugars such as lactose, fructose, galactose and pentoses.
  • CAP / CLSI formatted procedures available via fax or email - just add your name!
  • Store at Room Temperature

Specifications

Content And Storage 15°C to 30°C
Detectable Analytes Glucose, Lactose, Fructose, Galactose, Pentose
Sample Type Urine, Wine, Stool, Other Specimens
Disposable Single-use
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.