Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ AIN-76 Semipurified Diet, Pellet

Catalog No. MP290545320 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
10 kg
20 kg
50 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP290545320 20 kg
MP290545310 10 kg
MP290545350 50 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. MP290545320 Supplier MP Biomedicals™ Supplier No. 0290545320
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

A complete rodent diet for growth and maintenance during the first year of life in research labs.

Applications:
Ain-76 Semipurified Diet is a complete rodent diet for growth and maintenance during the first year of life in research labs. The diet has been found to be satisfactory for reproduction and lactation in both rats and mice.

Product Description:
Ain-76 Semipurified Diet is modified version of AIN-76 diet; (1) it has 10 fold increased amount of Vitamin-K; (2) contains an antioxidant.

TRUSTED_SUSTAINABILITY

Specifications

Product Type Animal Diet
Format Pellet
For Use With (Application) Basic animal diet
Quantity 20 kg
Storage Requirements 2°C to 8°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.