Learn More
Research Diets Inc RESEARCH DIETS INC Rodent Diet, AIN-76A, Purified, Irradiated
SDP
Catalog No. NC3980140
Encompass
This AIN-76A diet is a widely used, balanced, and purified rodent diet. It is suitable for various research areas including obesity, diabetes, osteoporosis, hypertension, atherosclerosis, and metabolic syndrome. This diet also serves as an effective control diet for studies.
  • Widely used, balanced, and purified rodent diet.
  • Supports research in obesity, diabetes, osteoporosis, hypertension, atherosclerosis, and metabolic syndrome.
  • Functions as a control diet in studies.
  • Packaged in a 12.5 kg box.
  • Gamma-irradiated for enhanced safety.
  • Available in pellet, powder, or liquid form.
  • Requires storage in a cool, dry environment for a shelf life of 3 to 6 months.
Catalog No. NC3980140 Supplier Research Diets Inc Supplier No. D10001I
This AIN-76A diet is a widely used, balanced, and purified rodent diet. It is suitable for various research areas including obesity, diabetes, osteoporosis, hypertension, atherosclerosis, and metabolic syndrome. This diet also serves as an effective control diet for studies.
  • Widely used, balanced, and purified rodent diet.
  • Supports research in obesity, diabetes, osteoporosis, hypertension, atherosclerosis, and metabolic syndrome.
  • Functions as a control diet in studies.
  • Packaged in a 12.5 kg box.
  • Gamma-irradiated for enhanced safety.
  • Available in pellet, powder, or liquid form.
  • Requires storage in a cool, dry environment for a shelf life of 3 to 6 months.