Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bio-Serv™ AIN-76A Rodent Diet
SDP

Catalog No. 14726614
Encompass
Change view
Click to view available options
Particle Shape:
Pellet
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Particle Shape
14-726-614 Pellet
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 14-726-614 Supplier Bio-Serv™ Supplier No. F1515
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chlorophyll free—does not interfere with imaging studies

  • Purified and extensively used in research
  • Balanced to meet nutritional needs of rodents
  • Phytoestrogen free
  • Nutritionally assayed
  • Ingredients: Sucrose, Dextrose, Casein, Corn Starch, Corn Oil, Cellulose, Mineral Mix, Vitamin Mix, DL-Methionine, Choline Bitartrate

Specifications

Product Type Rodent Diet
Components Sucrose, Casein, Corn Starch, Corn Oil, Cellulose, Mineral Mix, Vitamin Mix, DL-Methionine, Choline Bitartrate
For Use With (Application) Used in research
Particle Shape Pellet
Quantity 5000 g
Species Rodent
Diameter (English) 0.5 in.
Diameter (Metric) 1.27 cm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.