Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ AIN-76A Semipurified Diet

Catalog No. 0296009750 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
10 kg
20 kg
50 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
02-960-09750 50 kg
02-960-09710 10 kg
02-960-09720 20 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 02-960-09750 Supplier MP Biomedicals™ Supplier No. 0296009750
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

AIN-76A Semipurified Diet is intended for growth and maintenance of animals during the first year of life.

  • Contains casein purified high nitrogen, DL-methionine, sucrose, corn starch, Alphacel (non-nutritive bulk), corn oil, choline bitartrate, AIN mineral mix, and a 10-fold increase in the amount of vitamin K source to 500 μg/kg.
TRUSTED_SUSTAINABILITY

Specifications

Format Pellets
Quantity 50 kg
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.