Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bio-Serv™ AIN-93G Rodent Diet
SDP

Catalog No. 14726616
Encompass
Change view
Click to view available options
Format:
Pellet
Powder
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Format
14-726-616 Powder
14-726-530 Pellet
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 14-726-616 Supplier Bio-Serv™ Supplier No. F3197
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Purified diet formulated to meet the nutritional requirements of growing and gestating rodents

  • Widely used and published
  • Phytoestrogen free
  • Chlorophyll free—does not interfere with imaging studies
  • Nutritionally assayed
  • Ingredients: Corn Starch, Casein, Maltodextrin, Sucrose, Soybean Oil, Cellulose, Mineral Mix, Vitamin Mix, L-Cystine, Choline Bitartrate, tBHQ

Specifications

Product Type Rodent Diet
Components Corn Starch, Casein, Maltodextrin, Sucrose, Soybean Oil, Cellulose, Mineral Mix, Vitamin Mix, L-Cystine, Choline Bitartrate, tBHQ
Format Powder
For Use With (Application) Used and published for research objectives
Particle Shape Powder
Quantity 1 kg
Species Rodent
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.