Learn More
Enzo Life Sciences AIP (1mg)
SDP
Supplier:  Enzo Life Sciences BMLP2110001
Encompass_Preferred
AIP (autocamtide-2-related inhibitory peptide) is a highly specific and potent inhibitor of CaM kinase II. This peptide is derived from the CaM kinase II substrate autocamtide-2 (BML-P101) but alanine is substituted for threonine at the 9-position. AIP at 1 µM completely inhibited CaM kinase II but did not affect PKC, PKA, CaMKIV and other brain extract kinases. A cell-permeable, myristoylated version is available (BML-P212). Alternative name: Autocamtide 2 related inhibitory peptide. Purity: ≥97%. Long Term Storage: -20°C.
Catalog No. 50-201-1286