Learn More
Enzo Life Sciences Autocamtide-2-related inhibitory peptide (5mg). CAS: 167114-91-2
SDP
Supplier:  Enzo Life Sciences ALX151029M005
Encompass_Preferred
Highly specific and potent inhibitor of Ca2+/calmodulin-dependent protein kinase II without affecting cAMP-dependent protein kinase (PKA), protein kinase C (PKC) and Ca2+/calmodulin-dependent protein kinase IV. Alternative name: AIP. Formula: C64H116N22O19. MW: 1497.8. Purity: ≥98%. Appearance: Lyophilized. Solubility: Soluble in water. Long Term Storage: -20°C.
Catalog No. 50-200-7766