Learn More
Aviva Systems Biology Corp AIP Antibody - N-terminal region (ARP37219_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP37219_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against AIP. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide corresponding to a region of Mouse. Alternate names: Ara9, Xap2, Fkbp16, AA408703, AW476050, D19Bwg1412eApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rat, Zebrafish
Catalog No. 50-014-56338