Learn More
Aviva Systems Biology Corp AIP Antibody - middle region (ARP58131_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP58131_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against AIP. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the middle region of human AIP. Alternate names: ARA9, FKBP16, FKBP37, SMTPHN, XAP2, XAP-2ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rat
Catalog No. 50-286-442