Learn More
Aviva Systems Biology Corp Aip Antibody - middle region (ARP37218_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP37218_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Aip. It was validated on Western Blot. Aip,aryl-hydrocarbon receptor-interacting protein,Ara9; Xap2; Fkbp16; AA408703; AW476050; D19Bwg1412e; Aip,AH receptor-interacting protein,AH receptor-interacting protein,OTTMUSP00000032334,OTTMUSP00000032336The immunogen is a synthetic peptide corresponding to a region of Mouse. Alternate names: AA408703, AW476050, Ara9, D19Bwg1412e, Fkbp16, Xap2ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rat, Zebrafish
Catalog No. 50-284-599