Learn More
Carbosynth LLC. AIP-I (0.5 mg vial), CAS# 200010-29-3, MF= C43H60N8O13S2
SDP
Supplier:  Carbosynth LLC. AIP4515V0.5MG
Encompass_Preferred
Autoinducing Peptide-I; AIP1; Agr D1 Thiolactone; AP-1; Tyr-Ser-Thr-Cys-Asp-Phe-Ile-Met , Application: A Peptide Modulator of Quorum Sensing in Staphylococcus spp., Notes: Thioester bond between Cys4-Met8, salt: Trifluoroacetate Salt, origin/species: Staphylococcus aureus, one letter code: YSTCDFIM, MW: 961.11
Catalog No. 50-195-2391