Learn More
Carbosynth LLC. AIP-II (0.5 mg vial), CAS# 200010-31-7, MF= C38H58N10O12S
SDP
Supplier:  Carbosynth LLC. AIP4516V0.5MG
Encompass_Preferred
Autoinducing Peptide-II, AIP2; AIP-2; Gly-Val-Asn-Ala-Cys-Ser-Ser-Leu-Phe , Application: A Peptide Modulator of Quorum Sensing in Staphylococcus spp., Notes: Thioester bond between Cys5-Phe9, salt: Trifluoroacetate Salt, origin/species: Staphylococcus aureus, one letter code: GVNACSSLF , MW: 878.99
Catalog No. 50-195-2392