Learn More
Enzo Life Sciences AIP (myristoylated) (0.5mg)
SDP
Supplier:  Enzo Life Sciences BMLP2120500
Encompass_Preferred
Alternative name: Autocamtide 2 related inhibitory peptide (myristoylated). Purity: ≥95% (HPLC). Formulation: Lyophilized. Trifluoroacetate salt. Solubility: Soluble in 30% Acetic Acid in Water (5 mg/ml). Long Term Storage: -20°C.
Catalog No. 50-201-1287