Learn More
Enzo Life Sciences Autocamtide-2-related inhibitory peptide (myristoylated) (0.5mg)
SDP
Supplier:  Enzo Life Sciences ALX151030MC05
Encompass_Preferred
Cell permeable CaM kinase II inhibitor. Corresponds to ALX-151-029 but this one is N-terminal myristoylated to increase cell permeability. Alternative name: AIP (myristoylated). Formula: C78H142N22O20. MW: 1708.1. Purity: ≥98%. Appearance: Lyophilized. Solubility: Soluble in water. Long Term Storage: -20°C.
Catalog No. 50-200-7767