Learn More
Bachem Autocamtide-2-Related Inhibitory Peptide Trifluoroacetate
SDP
Supplier:  Bachem 4026616.0001
Encompass_Preferred
Autocamtide-2-Related Inhibitory Peptide Trifluoroacetate, H-Lys-Lys-Ala-Leu-Arg-Arg-Gln-Glu-Ala-Val-Asp-Ala-Leu-OH, 167114-91-2, C64H116N22O19, Mr 1497.76, 1mg. AIP, (Ala9)-Autocamtide-2. AIP, non-phosphorylatable analog of autocamtide-2, was found to be a highly specific and potent inhibitor of calmodulin-dependent protein Kinase II (IC50= 40 nM) without affecting cyclic AMP-dependent protein kinase, protein kinase C and calmodulin-dependent protein kinase IV.
Catalog No. 50-194-7008