Learn More
Sigma Aldrich Air Adaptor For 3 Ml Cartridge
Catalog No. 9180161EA Shop All MilliporeSigma Sigma Organic Chemistry Products
Click to view available options
Quantity:
1 Ea.
Catalog No. Quantity
91-801-61EA 1 Ea.
1 options
Catalog No. 91-801-61EA Supplier Sigma Aldrich Supplier No. 9180161EA
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.
The air adaptor connects cartridge to the air output of a fluid dispenser unit, such as a 3D bioprinter. Each adapter assembly is precision molded for secure attachment to the cartridge with minimal effort, making its connection to the air output of a fluid dispensing unit fast and seamless
Specifications
Quantity 1 Ea.