Learn More
Chemglass Life Sciences Air Aeration Tube for 15L Bio Reactors
SDP
Catalog No. 501294466
Encompass
Air Aeration Tube for 15L Bio Reactors. This tube is to be used when high gas flow rates are required. The sparger pipe extends to the bottom of the vessel and terminates just below the impellers on the stirrer shaft. This allows bubbles coming out of the sparger to rise up into the impellers where they are broken up for improved oxygen transfer.Like-for-like replacement parts for Applikon Biotechnology and Broadley-James brands of bioreactors.Applikon is a registered trademark of Applikon Biotechnology, Inc. Broadley-James and Broadley-James Corporation are registered trademarks of Broadley-James Corporation.
Catalog No. 50-129-4466 Supplier Chemglass Life Sciences Supplier No. CLS138518
Air Aeration Tube for 15L Bio Reactors. This tube is to be used when high gas flow rates are required. The sparger pipe extends to the bottom of the vessel and terminates just below the impellers on the stirrer shaft. This allows bubbles coming out of the sparger to rise up into the impellers where they are broken up for improved oxygen transfer.Like-for-like replacement parts for Applikon Biotechnology and Broadley-James brands of bioreactors.Applikon is a registered trademark of Applikon Biotechnology, Inc. Broadley-James and Broadley-James Corporation are registered trademarks of Broadley-James Corporation.