Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Grainger Air Amplifier Kit 5 In Inlet 50 CFM

Supplier:  Grainger 15J065

Encompass_Preferred

Air Amplifier Kit 5 In Inlet 50 CFM

Catalog No. 50-275-5759


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.