Learn More
Am Bickford Omnicon F/AIR Anesthesia Gas Filter 
SDP
Catalog No. NC0111395
Encompass_Preferred
Omnicon F/aIR cannister unit is a sensible answer to anesthesia gas problems in the operating room, the F/aIR anesthesia gas filter was specifically designed to remove waste anesthesia gases such as Isoflurane, Halothane, Enflurane and Sevoflurane, etc. from the operating room environment. Economical to use, with an average useful life of 12 to 15 hours, the F/aIR is easily adaptable to anesthesia machines. The F/aIR can remove no less than 50 grams of pure halogenated anesthetic gases and can then be conveniently discarded.
This item is not returnable. View return policy
Catalog No. NC0111395 Supplier Am Bickford Supplier No. 80120
Omnicon F/aIR cannister unit is a sensible answer to anesthesia gas problems in the operating room, the F/aIR anesthesia gas filter was specifically designed to remove waste anesthesia gases such as Isoflurane, Halothane, Enflurane and Sevoflurane, etc. from the operating room environment. Economical to use, with an average useful life of 12 to 15 hours, the F/aIR is easily adaptable to anesthesia machines. The F/aIR can remove no less than 50 grams of pure halogenated anesthetic gases and can then be conveniently discarded.