Learn More
Anton Paar Air Cooling Module for DSC
Catalog No. 5000580454
Encompass_Preferred
Air Cooling Module for Julia DSC x00A unique and patented cooling solution by Anton Paar.Powerful Peltier technology enables reaching sub-zerotemperatures and high cooling rates without a need for anexternal cooling device.Temperature range: -35°C to 700°CMax. heating rate: 300 K/minMax. cooling rate: 150 K/minCooling 10 K/min down to: 5°CCooling 5 K/min down to: -15°C
Catalog No. 50-005-80454 Supplier Anton Paar Supplier No. 261471
Air Cooling Module for Julia DSC x00A unique and patented cooling solution by Anton Paar.Powerful Peltier technology enables reaching sub-zerotemperatures and high cooling rates without a need for anexternal cooling device.Temperature range: -35°C to 700°CMax. heating rate: 300 K/minMax. cooling rate: 150 K/minCooling 10 K/min down to: 5°CCooling 5 K/min down to: -15°C