Learn More
Meridian Life Science Air-Dryable Direct DNA LAMP, 1ML
SDP
Supplier:  Meridian Life Science MDX1361
Encompass_Preferred
Air-Dryable Direct DNA LAMP Saliva mix combines even greater tolerance with fast, robust isothermal amplification to enable highly sensitive direct detection using crudely processed saliva or sputum samples. The mix is uniquely designed to overcome much higher concentrations of inhibitors, and so do not require a DNA purification step to concentrate and purify a patient sample. Very fast reaction kinetics result in faster time to results and superior amplification sensitivity and specificity. In addition, the 4x concentrated master mix is formulated with air-drying excipients which are optimized for oven-drying to create ambient-temperature stable assays, making it ideal for POC saliva testing.
Catalog No. 50-008-56542