Learn More
Meridian Life Science Air-Dryable Direct DNA qPCR, 1ML
Supplier: Meridian Life Science MDX1401
Air-Dryable DNA qPCR Stool is a 4x qPCR master mix for the development of DNA stool tests, enabling highly sensitive detection of target DNA from crudely processed stool samples. The mix has been formulated specifically to overcome the inhibitors found in stool samples – no further optimization is required aside from the addition of primers and probes. Furthermore, these mixes can be used in a wet format or oven dried to create ambient-temperature stable tests.