Learn More
Meridian Life Science Air-Dryable Direct DNA qPCR, 50ML
Supplier: Meridian Life Science MDX09250A
Air-Dryable Direct DNA qPCR Blood, 4x Buffer is a combination of the latest advances in buffer chemistry together with enhancers, stabilizers, dNTPs and MgCl2 at optimal concentrations for the development of DNA blood tests, enabling highly sensitive detection of target DNA from whole blood, plasma, and serum samples. The mix has been formulated specifically to overcome the inhibitor found in blood samples.