Learn More
Meridian Life Science Air-Dryable Direct DNA qPCR, 50ML
SDP
Supplier:  Meridian Life Science MDX09250
Encompass_Preferred
Air-Dryable DNA qPCR Blood is a 4x qPCR master mix for the development of DNA blood tests, enabling highly sensitive detection of target DNA from whole blood, plasma, and serum samples. The mix has been formulated specifically to overcome the inhibitors found in blood samples – no further optimization is required aside from the addition of primers and probes. Furthermore, these mixes can be used in a wet format or oven dried to create ambient-temperature stable whole blood tests, plasma tests or serum tests.
Catalog No. 50-008-56556