Learn More
Meridian Life Science Air-Dryable Direct RNA/DNA, 5ML
SDP
Supplier:  Meridian Life Science MDX1315
Encompass_Preferred
Air-Dryable RNA/DNA qPCR Saliva is a 4x qPCR master mix for the development of both DNA and RNA saliva tests from crudely processed saliva or sputum samples. The mix has been formulated specifically to overcome the inhibitors found in saliva samples – no further optimization is required aside from the addition of primers and probes. Furthermore, these mixes can be used in a wet format or oven dried to create ambient-temperature stable tests.
Catalog No. 50-008-56325