Learn More
Meridian Life Science Air-Dryable Direct RNA/DNA, 5ML
Supplier: Meridian Life Science MDX1415
Air-Dryable RNA/DNA qPCR Stool is a 4x qPCR master mix for the development of both DNA and RNA stool tests from crudely processed stool samples. The mix has been formulated specifically to overcome the inhibitors found in stool samples – no further optimization is required aside from the addition of primers and probes. Furthermore, these mixes can be used in a wet format or oven dried to create ambient-temperature stable tests.