Learn More
Meridian Life Science Air-Dryable Direct RNA/DNA, 5ML
SDP
Supplier:  Meridian Life Science MDX1215
Encompass_Preferred
Air-Dryable RNA/DNA qPCR Blood is a 4x qPCR master mix for the development of both DNA and RNA blood tests from whole blood, plasma, and serum samples. The mix has been formulated specifically to overcome the inhibitors found in blood samples – no further optimization is required aside from the addition of primers and probes. Furthermore, these mixes can be used in a wet format or oven dried to create ambient-temperature stable whole blood tests, plasma tests or serum tests.
Catalog No. 50-008-56335