Learn More
Meridian Life Science Air-Dryable qPCR Mix 4x, 50ML
SDP
Catalog No. 5000856343
Encompass_Preferred
Air-Dryable qPCR Mix (4x) is a glycerol-free qPCR mix containing an antibody-mediated hot-start Taq polymerase, reaction buffer, dNTPs, MgCl2 and air-dry compatible excipients. It is used for the development of multiplex molecular diagnostic tests, enabling highly sensitive detection of DNA targets. The air drying does not affect the specificity or reproducibility of the assay but increases the flexibility of the volume of sample that can be added, increasing sensitivity and also makes it ideal for microfluidic qPCR and point of care platforms.
Catalog No. 50-008-56343 Supplier Meridian Life Science Supplier No. MDX08250
Air-Dryable qPCR Mix (4x) is a glycerol-free qPCR mix containing an antibody-mediated hot-start Taq polymerase, reaction buffer, dNTPs, MgCl2 and air-dry compatible excipients. It is used for the development of multiplex molecular diagnostic tests, enabling highly sensitive detection of DNA targets. The air drying does not affect the specificity or reproducibility of the assay but increases the flexibility of the volume of sample that can be added, increasing sensitivity and also makes it ideal for microfluidic qPCR and point of care platforms.