Learn More
Meridian Life Science Air-Dryable RT-LAMP 1-Step, 1ML
SDP
Catalog No. 5000856308
Encompass_Preferred
Air-Dryable RT-LAMP 1-Step Mix is a glycerol-free mix for isothermal applications such as Loop-Mediated Isothermal Amplification (LAMP). It contains Reverse Transcriptase, Bst-DNA Polymerase (exo-), reaction buffer, dNTP and provided with air-dryable excipients. This is a robust master mix for loop-mediated isothermal amplification (LAMP) reactions, has been designed for amplification of RNA and DNA targets. In order to produce dried, ambient temperature stable RT-LAMP assays, assay specific primers are added to Air-Dryable RNA/DNA LAMP for subsequent air-drying.
Catalog No. 50-008-56308 Supplier Meridian Life Science Supplier No. MDX1181
Air-Dryable RT-LAMP 1-Step Mix is a glycerol-free mix for isothermal applications such as Loop-Mediated Isothermal Amplification (LAMP). It contains Reverse Transcriptase, Bst-DNA Polymerase (exo-), reaction buffer, dNTP and provided with air-dryable excipients. This is a robust master mix for loop-mediated isothermal amplification (LAMP) reactions, has been designed for amplification of RNA and DNA targets. In order to produce dried, ambient temperature stable RT-LAMP assays, assay specific primers are added to Air-Dryable RNA/DNA LAMP for subsequent air-drying.