Learn More
MSC AIR FLTR,MERV 9,12X24X1,MULT24
Catalog No. NC2076556
Encompass
** Per supplier requirements this item must be ordered in multiples of 24**12 x 24 x 1", MERV 9, 35% Efficiency, Panel Air FilterLofted Synthetic Polyester Fiber, Galvanized Steel Frame, 300 Max FPM, 600 CFM
Catalog No. NC2076556 Supplier MSC Supplier No. 43803741
** Per supplier requirements this item must be ordered in multiples of 24**12 x 24 x 1", MERV 9, 35% Efficiency, Panel Air FilterLofted Synthetic Polyester Fiber, Galvanized Steel Frame, 300 Max FPM, 600 CFM